Få mere at vide
Diethylene Glycol Dibenzoate, TRC
Artikelnummer. 17908214
Skift visning
Mængde:
100 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17908214 100 g
17937834 500 g
17969844 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17908214 Leverandør Toronto Research Chemicals Leverandørnr. TRCD444345100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 120-55-8
Molekylær formel C18 H18 O5
Molekylvægt (g/mol) 314.33
Synonym Diethylene glycol, dibenzoate (6CI,7CI,8CI), Ethanol, 2,2'-oxybis-, dibenzoate (9CI), 2-[2-(Benzoyloxy)ethoxy]ethyl benzoate, Benzoflex 2-42, Benzoflex 2-45, Benzoflex S 45, DEDB, Di-O-benzoyldiethylene glycol, Flexol 2GB, K-Flex DE, Monocizer PB 3, Monocizer PB 3A, PB 3, PB 3A, Velsicol 2-45, Diethylene Glycol Dibenzoate
IUPAC navn 2-(2-benzoyloxyethoxy)ethyl benzoate
SMIL O=C(OCCOCCOC(=O)c1ccccc1)c2ccccc2
Tekniske data
Kemisk navn eller materiale Diethylene Glycol Dibenzoate
Mængde 100 g
InChI formel InChI=1S/C18H18O5/c19-17(15-7-3-1-4-8-15)22-13-11-21-12-14-23-18(20)16-9-5-2-6-10-16/h1-10H,11-14H2
Formel vægt 314.1154