Tekniske data
| Oversigt | White to off-white |
|---|---|
| Indhold og opbevaring | 2°C to 8°C |
| Format | Powder |
| CAS | 364-98-7 |
| Molekylær formel | C8H7ClN2O2S |
| Formel vægt | 230.67 |
| Synonym | 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide; |
| Smeltepunkt | 330°C to 331°C |
| Opløselighed | Soluble in 0.1M NaOH. Insoluble in water or in methanol. |
| Assay | 98% |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
15467199
|
Thermo Scientific™
J66010.ME |
500 mg |
1298.53 DKK
500mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15457199
|
Thermo Scientific™
J66010.03 |
1 g |
2208.99 DKK
1g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Tekniske data
| White to off-white | |
| Powder | |
| C8H7ClN2O2S | |
| 7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide; | |
| Soluble in 0.1M NaOH. Insoluble in water or in methanol. |
| 2°C to 8°C | |
| 364-98-7 | |
| 230.67 | |
| 330°C to 331°C | |
| 98% |
Sikkerhed og håndtering
missing translation for 'einecsNumber' : 206-668-1
Anbefalet opbevaring : Keep cold