Få mere at vide
Desloratadine Epoxide, TRC
Artikelnummer. 30353155
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30353155 25 mg -
30359986 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30353155 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29027525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1346604-23-6
Molekylær formel C19H19ClN2O
Molekylvægt (g/mol) 326.82
Synonym Epoxy Desloratadine
SMIL Clc1ccc2c(CCc3cccnc3C24OC45CCNCC5)c1
Tekniske data
Kemisk navn eller materiale Desloratadine Epoxide
Mængde 25 mg
InChI formel InChI=1S/C19H19ClN2O/c20-15-5-6-16-14(12-15)4-3-13-2-1-9-22-17(13)19(16)18(23-19)7-10-21-11-8-18/h1-2,5-6,9,12,21H,3-4,7-8,10-11H2
Formel vægt 326.1186