Få mere at vide
Desethylcarbamoyl Cabergoline, TRC
Artikelnummer. 30360714
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30360714 10 mg
30358685 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30360714 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28886010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85329-86-8
Molekylær formel C23 H32 N4 O
Molekylvægt (g/mol) 380.53
Synonym (8β)-N-[3-(dimethylamino)propyl]-6-(2-propen-1-yl)ergoline-8-carboxamide, FCE 21590, (6aR,9R,10aR)-N-[3-(Dimethylamino)propyl]-7-(prop-2-enyl)-4,6,6a,7,8,9,10,10a-octahydroindolo[4,3-fg]quinoline-9-carboxamide, Cabergoline Imp. D (EP)
IUPAC navn (6aR,9R,10aR)-N-[3-(dimethylamino)propyl]-7-prop-2-enyl-6,6a,8,9,10,10a-hexahydro-4H-indolo[4,3-fg]quinoline-9-carboxamide
SMIL CN(C)CCCNC(=O)[C@@H]1C[C@H]2[C@@H](Cc3c[nH]c4cccc2c34)N(CC=C)C1
Tekniske data
Kemisk navn eller materiale Desethylcarbamoyl Cabergoline
Mængde 10 mg
InChI formel InChI=1S/C23H32N4O/c1-4-10-27-15-17(23(28)24-9-6-11-26(2)3)12-19-18-7-5-8-20-22(18)16(14-25-20)13-21(19)27/h4-5,7-8,14,17,19,21,25H,1,6,9-13,15H2,2-3H3,(H,24,28)/t17-,19-,21-/m1/s1
Formel vægt 380.2576