Få mere at vide
Deschloroloperamide Hydrochloride, TRC
Artikelnummer. 30137754
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137754 100 mg
30008962 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137754 Leverandør Toronto Research Chemicals Leverandørnr. TRCD226605100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1134369-22-4
Molekylær formel C29H35ClN2O2
Molekylvægt (g/mol) 479.05
Synonym 4-Hydroxy-N,N-dimethyl-α,α,4-triphenyl-1-piperidinebutanamide Hydrochloride
IUPAC navn 4-(4-hydroxy-4-phenylpiperidin-1-yl)-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride
SMIL CC1Nc2c(CC1)cc(F)cc2
Tekniske data
Kemisk navn eller materiale Deschloroloperamide Hydrochloride
Mængde 100 mg
InChI formel InChI=1S/C29H34N2O2.ClH/c1-30(2)27(32)29(25-14-8-4-9-15-25,26-16-10-5-11-17-26)20-23-31-21-18-28(33,19-22-31)24-12-6-3-7-13-24;/h3-17,33H,18-23H2,1-2H3;1H