Få mere at vide
Deschloroacetylmetolachlor Propanol, TRC
Artikelnummer. 30362425
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362425 10 mg
30360206 100 mg
30351217 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30362425 Leverandør Toronto Research Chemicals Leverandørnr. TRCD35392510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61520-53-4
Molekylær formel C12 H19 N O
Molekylvægt (g/mol) 193.29
Synonym 2-[(2-Ethyl-6-methylphenyl)amino]-1-propanol, CGA 37913, Deschloroacetylmetolachlor propanol, N-(1-Hydroxy-2-propyl)-2-ethyl-6-methylaniline
IUPAC navn 2-(2-ethyl-6-methylanilino)propan-1-ol
SMIL CCc1cccc(C)c1NC(C)CO
Tekniske data
Kemisk navn eller materiale Metolachlor-des(chloroacetyl)-O-desmethyl
Mængde 10 mg
InChI formel InChI=1S/C12H19NO/c1-4-11-7-5-6-9(2)12(11)13-10(3)8-14/h5-7,10,13-14H,4,8H2,1-3H3
Formel vægt 193.1467