Få mere at vide
Desalkyl Ebastine, TRC
Artikelnummer. 30138427
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138427 50 mg
30007052 10 mg
30010852 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138427 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28877050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 58258-01-8
Molekylær formel C18 H21 N O
Molekylvægt (g/mol) 267.37
Synonym 4-(Diphenylmethoxy)piperidine, 4-(4-Benzhydryloxy)piperidine, 4-(Benzhydryloxy)piperidine, 4-(Diphenylmethoxy)piperidine, Ebastine Imp. C (EP)
IUPAC navn 4-benzhydryloxypiperidine
SMIL C1CC(CCN1)OC(c2ccccc2)c3ccccc3
Tekniske data
Kemisk navn eller materiale Desalkyl Ebastine
Mængde 50 mg
InChI formel InChI=1S/C18H21NO/c1-3-7-15(8-4-1)18(16-9-5-2-6-10-16)20-17-11-13-19-14-12-17/h1-10,17-19H,11-14H2
Formel vægt 267.1623