Få mere at vide
Desacetyl Bisacodyl, TRC
Artikelnummer. 17994335
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17994335 5 mg
17930595 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17994335 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2819455MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 72901-16-7
Molekylær formel C20 H17 N O3
Molekylvægt (g/mol) 319.35
Synonym 4-[(RS)-(4-Hydroxyphenyl)-(pyridin-2-yl)methyl]phenyl Acetate, Bisacodyl Imp. C (EP)
IUPAC navn [4-[(4-hydroxyphenyl)-pyridin-2-ylmethyl]phenyl] acetate
SMIL CC(=O)Oc1ccc(cc1)C(c2ccc(O)cc2)c3ccccn3
Tekniske data
Kemisk navn eller materiale 4-[(RS)-(4-Hydroxyphenyl)-(pyridin-2-yl)methyl]phenyl Acetate
Mængde 5 mg
InChI formel InChI=1S/C20H17NO3/c1-14(22)24-18-11-7-16(8-12-18)20(19-4-2-3-13-21-19)15-5-9-17(23)10-6-15/h2-13,20,23H,1H3
Formel vægt 319.1208