Få mere at vide
Des(N,N-Dimethylprop-2-enyl-1-Amine) Afatinib, TRC
Artikelnummer. 30363480
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30363480 50 mg
30354688 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30363480 Leverandør Toronto Research Chemicals Leverandørnr. TRCD29124050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2223677-60-7
Molekylær formel C19H16ClFN4O3
Molekylvægt (g/mol) 402.81
Synonym Des(4-dimethylamino-2-en-1-oxo)butyl Afatinib N6 Formyl
IUPAC navn (R)-N-(4-((3-chloro-4-fluorophenyl)amino)-7-((tetrahydrofuran-3-yl)oxy)quinazolin-6-yl)formamide
SMIL FC(C=C1)=C(Cl)C=C1NC2=NC=NC3=CC(O[C@H]4COCC4)=C(NC([H])=O)C=C32
Tekniske data
Kemisk navn eller materiale Des(N,N-Dimethylprop-2-enyl-1-Amine) Afatinib
Mængde 50 mg
InChI formel InChI=1S/C19H16ClFN4O3/c20-14-5-11(1-2-15(14)21)25-19-13-6-17(24-10-26)18(7-16(13)22-9-23-19)28-12-3-4-27-8-12/h1-2,5-7,9-10,12H,3-4,8H2,(H,24,26)(H,22,23,25)/t12-/m1/s1
Formel vægt 402.09