Få mere at vide
Des-(N-2-pyridyl-Beta-alanine Ethyl Ester) Dabigatran Etexilate 5-Ethyl Carboxylate (Dabigatran Impurity), TRC
Artikelnummer. 30132765
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132765 5 mg
30008602 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132765 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2932755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1408238-36-7
Molekylær formel C26H33N5O4
Molekylvægt (g/mol) 479.57
Synonym 2-[[[4-[[[(Hexyloxy)carbonyl]amino]iminomethyl]phenyl]amino]methyl]-1-methyl-1H-benzimidazole-5-carboxylic Acid Ethyl Ester
SMIL CCCCCCOC(=O)NC(=N)c1ccc(NCc2nc3cc(ccc3n2C)C(=O)OCC)cc1
Tekniske data
Kemisk navn eller materiale Des-(N-2-pyridyl-beta-alanine Ethyl Ester) Dabigatran Etexilate 5-Ethyl Carboxylate (Dabigatran Impurity)
Mængde 5 mg
Formel vægt 479.2533