Få mere at vide
Des(1-cyclohexanol) Venlafaxine-d6, TRC
Artikelnummer. 30360751
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30360751 25 mg -
30365172 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30360751 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28859225MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1330046-00-8
Molekylær formel C11H11D6NO
Molekylvægt (g/mol) 185.3
Synonym 4-Methoxy-N,N-(dimethyl-d6)benzeneethanamine, p-Methoxy-N,N-(dimethyl-d6) phenethylamine, N,N-(Dimethyl-d6)-2-(p-anisyl)ethylamine, N,N-(Dimethyl-d6)-4-methoxyphenethylamine
SMIL COC1=CC=C(CCN(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C1
Tekniske data
Kemisk navn eller materiale Des(1-cyclohexanol) Venlafaxine-d6
Mængde 25 mg
InChI formel InChI=1S/C11H17NO/c1-12(2)9-8-10-4-6-11(13-3)7-5-10/h4-7H,8-9H2,1-3H3/i1D3,2D3