missing translation for 'onlineSavingsMsg'
Få mere at vide

Dehydropinaverium Bromide, TRC

Product Code. 30006308
Change view
Click to view available options
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30006308 25 mg
30004917 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006308 Supplier Toronto Research Chemicals Supplier No. TRCB68379525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1235355-01-7
Molekylær formel C26 H39 Br N O4 . Br
Molekylvægt (g/mol) 589.4
Synonym (1R,5S)-4-[(2-Bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl)ethoxy]ethyl]morpholinium Bromide (1:1), Morpholinium, 4-[(2-bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl)ethoxy]ethyl]-, bromide (1:1)
IUPAC navn 4-[(2-bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-[(1R,5S)-6,6-dimethyl-4-bicyclo[3.1.1]hept-3-enyl]ethoxy]ethyl]morpholin-4-ium;bromide
SMIL [Br-].COc1cc(Br)c(C[N+]2(CCOCCC3=CC[C@@H]4C[C@H]3C4(C)C)CCOCC2)cc1OC

Specifications

Kemisk navn eller materiale (1R,5S)-4-[(2-Bromo-4,5-dimethoxyphenyl)methyl]-4-[2-[2-(6,6-dimethylbicyclo[3.1.1]hept-2-en-2-yl)ethoxy]ethyl]morpholinium Bromide
Mængde 25 mg
InChI formel InChI=1S/C26H39BrNO4.BrH/c1-26(2)21-6-5-19(22(26)16-21)7-11-31-12-8-28(9-13-32-14-10-28)18-20-15-24(29-3)25(30-4)17-23(20)27;/h5,15,17,21-22H,6-14,16,18H2,1-4H3;1H/q+1;/p-1/t21-,22-;/m1./s1
Formel vægt 587.1246
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.