Få mere at vide
Dehydro Lercanidipine, TRC
Artikelnummer. 17991905
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17991905 100 mg
17913625 10 mg
17923295 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17991905 Leverandør Toronto Research Chemicals Leverandørnr. TRCD229220100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 887769-34-8
Molekylær formel C36 H39 N3 O6
Molekylvægt (g/mol) 609.71
Synonym 2,6-dimethyl-4-(3-nitrophenyl)-3,5-Pyridinedicarboxylic acid 3-[2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl] 5-methyl ester, 3,5-Pyridinedicarboxylic acid, 2,6-dimethyl-4-(3-nitrophenyl)-, 2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl methyl ester (9CI), Dehydro Lercanidipine, Dehydrolercanidipine
IUPAC navn 3-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 5-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate
SMIL COC(=O)c1c(C)nc(C)c(C(=O)OC(C)(C)CN(C)CCC(c2ccccc2)c3ccccc3)c1c4cccc(c4)[N+](=O)[O-]
Tekniske data
Kemisk navn eller materiale Dehydro Lercanidipine
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C36H39N3O6/c1-24-31(34(40)44-6)33(28-18-13-19-29(22-28)39(42)43)32(25(2)37-24)35(41)45-36(3,4)23-38(5)21-20-30(26-14-9-7-10-15-26)27-16-11-8-12-17-27/h7-19,22,30H,20-21,23H2,1-6H3
Formel vægt 609.2839
Procent renhed >95%