Få mere at vide
Defluoro Flunarizine Dihydrochloride, TRC
Artikelnummer. 30364546
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30364546 500 mg
30363986 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30364546 Leverandør Toronto Research Chemicals Leverandørnr. TRCD228700500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27064-96-6
Molekylær formel C26 H27 F N2 . 2 Cl H
Molekylvægt (g/mol) 459.43
Synonym Piperazine, 1-[(4-fluorophenyl)phenylmethyl]-4-(3-phenyl-2-propen-1-yl)-, hydrochloride (1:2), Piperazine, 1-[(4-fluorophenyl)phenylmethyl]-4-(3-phenyl-2-propenyl)-, dihydrochloride (9CI), Piperazine, 1-cinnamyl-4-(p-fluoro-α-phenylbenzyl)-, dihydrochloride (8CI), 1-[(RS)-(4-Fluorophenyl)phenylmethyl]-4-[(2EZ)-3-phenylprop-2-enyl]piperazine dihydrochloride, (EZ)-Defluoro-flunarizine dihydrochloride
IUPAC navn 1-[(4-fluorophenyl)-phenylmethyl]-4-(3-phenylprop-2-enyl)piperazine;dihydrochloride
SMIL Cl.Cl.Fc1ccc(cc1)C(N2CCN(CC=Cc3ccccc3)CC2)c4ccccc4
Tekniske data
Kemisk navn eller materiale Defluoro Flunarizine dihydrochloride
Mængde 500 mg
InChI formel InChI=1S/C26H27FN2.2ClH/c27-25-15-13-24(14-16-25)26(23-11-5-2-6-12-23)29-20-18-28(19-21-29)17-7-10-22-8-3-1-4-9-22;;/h1-16,26H,17-21H2;2*1H
Formel vægt 458.1692