CAS RN 27064-96-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Defluoro Flunarizine Dihydrochloride, TRC
CAS: 27064-96-6 Molekylær formel: C26 H27 F N2 . 2 Cl H Molekylvægt (g/mol): 459.43 Synonym: Piperazine, 1-[(4-fluorophenyl)phenylmethyl]-4-(3-phenyl-2-propen-1-yl)-, hydrochloride (1:2),Piperazine, 1-[(4-fluorophenyl)phenylmethyl]-4-(3-phenyl-2-propenyl)-, dihydrochloride (9CI),Piperazine, 1-cinnamyl-4-(p-fluoro-α-phenylbenzyl)-, dihydrochloride (8CI),1-[(RS)-(4-Fluorophenyl)phenylmethyl]-4-[(2EZ)-3-phenylprop-2-enyl]piperazine dihydrochloride,(EZ)-Defluoro-flunarizine dihydrochloride IUPAC navn: 1-[(4-fluorophenyl)-phenylmethyl]-4-(3-phenylprop-2-enyl)piperazine;dihydrochloride SMIL: Cl.Cl.Fc1ccc(cc1)C(N2CCN(CC=Cc3ccccc3)CC2)c4ccccc4