Få mere at vide
Defluoro Atorvastatin Acetonide tert-Butyl Ester, TRC
Artikelnummer. 30350448
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30350448 5 mg
30366566 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30350448 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2290005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1105067-91-1
Molekylær formel C40H48N2O5
Molekylvægt (g/mol) 636.82
Synonym (4R,6R)-2,2-Dimethyl-6-[2-[2-(1-methylethyl)-4,5-diphenyl-3-[(phenylamino)carbonyl]-1H-pyrrol-1-yl]ethyl]-1,3-dioxane-4-acetic Acid 1,1-Dimethylethyl Ester
IUPAC navn tert-butyl 2-[(4R,6R)-6-[2-[2,3-diphenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-dioxan-4-yl]acetate
SMIL CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccccc4)n1CC[C@@H]5C[C@H](CC(=O)OC(C)(C)C)OC(C)(C)O5
Tekniske data
Kemisk navn eller materiale Defluoro Atorvastatin Acetonide tert-Butyl Ester
Mængde 5 mg
InChI formel InChI=1S/C40H48N2O5/c1-27(2)36-35(38(44)41-30-21-15-10-16-22-30)34(28-17-11-8-12-18-28)37(29-19-13-9-14-20-29)42(36)24-23-31-25-32(46-40(6,7)45-31)26-33(43)47-39(3,4)5/h8-22,27,31-32H,23-26H2,1-7H3,(H,41,44)/t31-,32-/m1/s1