Få mere at vide
Decyl Octyl Phthalate, TRC
Artikelnummer. 30134722
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134722 500 mg
17943225 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134722 Leverandør Toronto Research Chemicals Leverandørnr. TRCD228460500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 119-07-3
Molekylær formel C26H42O4
Molekylvægt (g/mol) 418.61
Synonym 1,2-Benzenedicarboxylic acid, 1-decyl 2-octyl ester, 1,2-Benzenedicarboxylic acid, decyl octyl ester (9CI), Phthalic acid, decyl octyl ester (6CI,7CI,8CI), Decyl octyl phthalate, Dinopol 235, Octyl decyl phthalate, Polycizer 532, Polycizer 562, Staflex 500, n-Decyl n-octyl phthalate, n-Octyl n-decyl phthalate, Phthalic acid, octyldecyl ester
IUPAC navn 2-O-decyl 1-O-octyl benzene-1,2-dicarboxylate
SMIL CCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC
Tekniske data
Kemisk navn eller materiale Phthalic acid, octyldecyl ester
Mængde 500 mg
InChI formel InChI=1S/C26H42O4/c1-3-5-7-9-11-12-14-18-22-30-26(28)24-20-16-15-19-23(24)25(27)29-21-17-13-10-8-6-4-2/h15-16,19-20H,3-14,17-18,21-22H2,1-2H3
Formel vægt 418.308