CAS RN 119-07-3
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
Decyl Octyl Phthalate, TRC
CAS: 119-07-3 Molekylær formel: C26H42O4 Molekylvægt (g/mol): 418.61 Synonym: 1,2-Benzenedicarboxylic acid, 1-decyl 2-octyl ester,1,2-Benzenedicarboxylic acid, decyl octyl ester (9CI),Phthalic acid, decyl octyl ester (6CI,7CI,8CI),Decyl octyl phthalate,Dinopol 235,Octyl decyl phthalate,Polycizer 532,Polycizer 562,Staflex 500,n-Decyl n-octyl phthalate,n-Octyl n-decyl phthalate,Phthalic acid, octyldecyl ester IUPAC navn: 2-O-decyl 1-O-octyl benzene-1,2-dicarboxylate SMIL: CCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCC