Learn More
Borer Chemie™ deconex 21 LAB CLF
For automated cleaning of laboratory glassware and utensils. Thanks to its excellent material compatibility, deconex 21 LAB CLF also cleans sensitive objects made of aluminium. An acid rinsing with a deconex product is recommended.
Brand: Borer Chemie™ 500925.00-K10W
Description
- Concentrate for automated cleaning
- High yield, typical concentration: 5-10 mL/L
- Excellent material compatibility, also for aluminium
- Broad cleaning spectrum
- Can be rinsed off without leaving residues
- For consistent and flawless results
- Without chlorine, EDTA, NTA and surfactants
- Acid rinse with deconex cleaner recommended
- Technical support by the manufacturer
Specifications
Liquid | |
Alkalis, complexing agents, corrosion inhibitor, sequestering agents | |
Canister | |
1 x 12 kg | |
8.8 L | |
12 kg |
∼11.4 (1% Solution) | |
Concentrate | |
5°C to 25°C | |
2.3 gal. | |
26.5 lb. |
Safety and Handling
- deconex 21 LAB CLF
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : Tetrapotassiumdiphosphate, tripotassiumphosphate, dipotassium trioxosilicate