Learn More
Borer Chemie™ deconex 21 LAB CLF
For automated cleaning of laboratory glassware and utensils. Thanks to its excellent material compatibility, deconex 21 LAB CLF also cleans sensitive objects made of aluminium. An acid rinsing with a deconex product is recommended.
861.00 DKK - 4120.00 DKK
Specifications
Form | Liquid |
---|---|
pH | ∼11.4 (1% Solution) |
Composition | Alkalis, complexing agents, corrosion inhibitor, sequestering agents |
Concentration | Concentrate |
Container Type | Canister |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15430350
|
Borer Chemie™
500925.00-KK5W |
1 x 6 kg |
861.00 DKK
6kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17446492
|
Borer Chemie™
500925.00-K10W |
1 x 12 kg |
1570.00 DKK
12kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17456492
|
Borer Chemie™
500925.00-K25W |
1 x 32 kg |
4120.00 DKK
32kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Concentrate for automated cleaning
- High yield, typical concentration: 5-10 mL/L
- Excellent material compatibility, also for aluminium
- Broad cleaning spectrum
- Can be rinsed off without leaving residues
- For consistent and flawless results
- Without chlorine, EDTA, NTA and surfactants
- Acid rinse with deconex cleaner recommended
- Technical support by the manufacturer
Specifications
Liquid | |
Alkalis, complexing agents, corrosion inhibitor, sequestering agents | |
Canister |
∼11.4 (1% Solution) | |
Concentrate | |
5°C to 25°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- deconex 21 CLF
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : tetrapotassium pyrophosphate, potassiumsilicate, tripotassium phosphate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Borer Chemie™ deconex 21 LAB CLF