Få mere at vide
Decanoyl-10,10,10,d3-L-carnitine Chloride, TRC
Artikelnummer. 30361266
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361266 2.5 mg
30360032 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361266 Leverandør Toronto Research Chemicals Leverandørnr. TRCD2125822.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1297271-50-1
Molekylær formel C17H31D3ClNO4
Molekylvægt (g/mol) 354.93
Synonym (2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxodecyl)oxy]-1-propanaminium Chloride-d3
IUPAC navn (R)-3-carboxy-2-((decanoyl-10,10,10-d3)oxy)-N,N,N-trimethylpropan-1-aminium chloride
SMIL O=C(O[C@H](CC(O)=O)C[N+](C)(C)C)CCCCCCCCC([2H])([2H])[2H].[Cl-]
Tekniske data
Kemisk navn eller materiale Decanoyl-10,10,10,d3-L-carnitine Chloride
Mængde 2.5 mg
InChI formel InChI=1S/C17H33NO4.ClH/c1-5-6-7-8-9-10-11-12-17(21)22-15(13-16(19)20)14-18(2,3)4;/h15H,5-14H2,1-4H3;1H/t15-;/m1./s1/i1D3;
Formel vægt 354.23