Få mere at vide
Darifenacin Hydrobromide, TRC
Artikelnummer. 30011209
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011209 100 mg
30004119 25 mg
30007176 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011209 Leverandør Toronto Research Chemicals Leverandørnr. TRCD193400100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 133099-07-7
Molekylær formel C28 H30 N2 O2 . Br H
Molekylvægt (g/mol) 507.46
Synonym (3S)-1-[2-(2,3-Dihydro-5-benzofuranyl)ethyl]-α,α-diphenyl-3-pyrrolidineacetamide hydrobromide (1:1), 3-Pyrrolidineacetamide, 1-[2-(2,3-dihydro-5-benzofuranyl)ethyl]-α,α-diphenyl-, monohydrobromide, (3S)- (9CI), 3-Pyrrolidineacetamide, 1-[2-(2,3-dihydro-5-benzofuranyl)ethyl]-α,α-diphenyl-, monohydrobromide, (S)-, (S)-3-(Carbamoyldiphenylmethyl)-1-[2-(2,3-dihydrobenzofuran-5-yl)ethyl]pyrrolidine hydrobromide, Darifenacin hydrobromide, Darilong, Emselex, Enablex, UK 88525-04, UK 88525-04 hydrobromide
IUPAC navn 2-[(3S)-1-[2-(2,3-dihydro-1-benzofuran-5-yl)ethyl]pyrrolidin-3-yl]-2,2-diphenylacetamide;hydrobromide
SMIL Br.NC(=O)C([C@@H]1CCN(CCc2ccc3OCCc3c2)C1)(c4ccccc4)c5ccccc5
Tekniske data
Kemisk navn eller materiale Darifenacin Hydrobromide
Mængde 100 mg
InChI formel InChI=1S/C28H30N2O2.BrH/c29-27(31)28(23-7-3-1-4-8-23,24-9-5-2-6-10-24)25-14-17-30(20-25)16-13-21-11-12-26-22(19-21)15-18-32-26;/h1-12,19,25H,13-18,20H2,(H2,29,31);1H/t25-;/m1./s1
Formel vægt 506.1569