Få mere at vide
Dacthal Monoacid, TRC
Artikelnummer. 30006907
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006907 10 mg
30008486 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006907 Leverandør Toronto Research Chemicals Leverandørnr. TRCD10165010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 887-54-7
Molekylær formel C9 H4 Cl4 O4
Molekylvægt (g/mol) 317.94
Synonym 1,4-Benzenedicarboxylic acid, 2,3,5,6-tetrachloro-, monomethyl ester (9CI), Terephthalic acid, tetrachloro-, methyl ester (7CI), Terephthalic acid, tetrachloro-, monomethyl ester (8CI), Chlorthal monomethyl, Dacthal monoacid, Methyl 2,3,5,6-tetrachloroterephthalate, Monomethyl 2,3,5,6-tetrachloroterephthalate, Monomethyl tetrachloroterephthalate, Tetrachloroterephthalic acid methyl ester
IUPAC navn 2,3,5,6-tetrachloro-4-methoxycarbonylbenzoic acid
SMIL COC(=O)c1c(Cl)c(Cl)c(C(=O)O)c(Cl)c1Cl
Tekniske data
Kemisk navn eller materiale Dacthal monoacid
Mængde 10 mg
InChI formel InChI=1S/C9H4Cl4O4/c1-17-9(16)3-6(12)4(10)2(8(14)15)5(11)7(3)13/h1H3,(H,14,15)
Formel vægt 315.8864