Få mere at vide
Dacarbazine, Thermo Scientific™
1097.03 DKK - 1962.72 DKK
Kemiske identifikatorer
| CAS | 4342-03-4 |
|---|---|
| Molekylær formel | C4H3IN2O |
| Molekylvægt (g/mol) | 221.99 |
| MDL nummer | MFCD00057167 |
| InChI nøgle | SMIGOGPUTQXOKF-UHFFFAOYSA-N |
| Synonym | 5-iodopyrimidin-4-ol, 5-iodopyrimidin-4 3h-one, 4-hydroxy-5-iodopyrimidine, 5-iodo-1h-pyrimidin-4-one, 4 1h-pyrimidinone, 5-iodo, 5-iodopyrimidin-4 1h-one, 5-iodo-3h-pyrimidin-4-one, 5-iodo-3,4-dihydropyrimidin-4-one, 5-iodopyrimidin-4-ol;5-iodopyrimidin-4 1h-one |
| PubChem CID | 12356582 |
| IUPAC navn | 5-iodo-1H-pyrimidin-6-one |
| SMIL | C1=C(C(=O)NC=N1)I |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
15463689
|
Thermo Scientific Alfa Aesar
J61023.ME |
500mg |
1097.03 DKK
500mg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15453689
|
Thermo Scientific Alfa Aesar
J61023.03 |
1g |
1962.72 DKK
1g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Used in treatment of malignant melanoma and sarcomas
- Generates methyl adducts to purine bases in DNA
Kemiske identifikatorer
| C4H3IN2O | |
| MFCD00057167 | |
| 5-iodopyrimidin-4-ol, 5-iodopyrimidin-4 3h-one, 4-hydroxy-5-iodopyrimidine, 5-iodo-1h-pyrimidin-4-one, 4 1h-pyrimidinone, 5-iodo, 5-iodopyrimidin-4 1h-one, 5-iodo-3h-pyrimidin-4-one, 5-iodo-3,4-dihydropyrimidin-4-one, 5-iodopyrimidin-4-ol;5-iodopyrimidin-4 1h-one | |
| 5-iodo-1H-pyrimidin-6-one |
Tekniske data
| Dacarbazine | |
| C4H3IN2O | |
| 14,2798 | |
| Slightly soluble in water,DMSO and ethanol /n | |
| C1=C(C(=O)NC=N1)I | |
| 221.99 | |
| 182.18 |
| 4342-03-4 | |
| MFCD00057167 | |
| 5-iodopyrimidin-4-ol, 5-iodopyrimidin-4 3h-one, 4-hydroxy-5-iodopyrimidine, 5-iodo-1h-pyrimidin-4-one, 4 1h-pyrimidinone, 5-iodo, 5-iodopyrimidin-4 1h-one, 5-iodo-3h-pyrimidin-4-one, 5-iodo-3,4-dihydropyrimidin-4-one, 5-iodopyrimidin-4-ol;5-iodopyrimidin-4 1h-one | |
| SMIGOGPUTQXOKF-UHFFFAOYSA-N | |
| 5-iodo-1H-pyrimidin-6-one | |
| 12356582 |
Sikkerhed og håndtering
GHS H Statement
H311-H340-H350-H332-H315-H319-H335
Toxic in contact with skin.
May cause genetic defects.
May cause cancer.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P332+P313-P362-P501c
H312+H332-H315-H319-H335-H350
missing translation for 'einecsNumber' : 224-396-1
missing translation for 'rtecsNumber' : NI3950000
missing translation for 'tsca' : No
Anbefalet opbevaring : Keep cold
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.