Få mere at vide
Dacarbazine, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J61023.03
| Mængde | 1g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Used in treatment of malignant melanoma and sarcomas
- Generates methyl adducts to purine bases in DNA
Kemiske identifikatorer
| C6H10N6O | |
| MFCD00057167 | |
| dacarbazine, biocarbazine, deticene, dtic-dome, biocarbazine r, decarbazine, dtic, icdt, dacarbazinum inn-latin, dacarbazino inn-spanish | |
| (5E)-5-(dimethylaminohydrazinylidene)imidazole-4-carboxamide |
Tekniske data
| Dacarbazine | |
| C6H10N6O | |
| 1g | |
| dacarbazine, biocarbazine, deticene, dtic-dome, biocarbazine r, decarbazine, dtic, icdt, dacarbazinum inn-latin, dacarbazino inn-spanish | |
| OMJKFYKNWZZKTK-UXBLZVDNSA-N | |
| (5E)-5-(dimethylaminohydrazinylidene)imidazole-4-carboxamide | |
| 5353562 |
| 4342-03-4 | |
| MFCD00057167 | |
| 14,2798 | |
| Slightly soluble in water,DMSO and ethanol /n | |
| CN(C)NN=C1C(=NC=N1)C(=O)N | |
| 182.19 | |
| 182.18 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.