Få mere at vide
Dabigatran-13C6, TRC
Artikelnummer. 30133490
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133490 10 mg
30132825 5 mg
17951525 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133490 Leverandør Toronto Research Chemicals Leverandørnr. TRCD10009110MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1210608-88-0
Molekylær formel C1913C6H25N7O3
Molekylvægt (g/mol) 477.47
Synonym N-[[2-[[[4-(Aminoiminomethyl)phenyl]amino]methyl]-1-methyl-1H-benzimidazol-5-yl]carbonyl]-N-2-pyridinyl-β-alanine-13C6, BIBR 953-13C6, BIBR 953ZW-13C6
IUPAC navn 3-[[2-[[(4-carbamimidoyl(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-trien-1-yl)amino]methyl]-1-methylbenzimidazole-5-carbonyl]-pyridin-2-ylamino]propanoic acid
SMIL O=C(N(C1=CC=CC=N1)CCC(O)=O)C2=CC=C3N(C(CN[13C]4=[13CH][13CH]=[13C](C(N)=N)[13CH]=[13CH]4)=NC3=C2)C
Tekniske data
Kemisk navn eller materiale Dabigatran-13C6
Mængde 10 mg
InChI formel InChI=1S/C25H25N7O3/c1-31-20-10-7-17(25(35)32(13-11-23(33)34)21-4-2-3-12-28-21)14-19(20)30-22(31)15-29-18-8-5-16(6-9-18)24(26)27/h2-10,12,14,29H,11,13,15H2,1H3,(H3,26,27)(H,33,34)/i5+1,6+1,8+1,9+1,16+1,18+1
Formel vægt 477.222