Få mere at vide
D-Tetrahydropalmatine, TRC
Artikelnummer. 30418865
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418865 100 mg
30422567 25 mg
30416888 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30418865 Leverandør Toronto Research Chemicals Leverandørnr. TRCT294160100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3520-14-7
Molekylær formel C21H25NO4
Molekylvægt (g/mol) 355.43
Synonym (13aR)-5,8,13,13a-Tetrahydro-2,3,9,10-tetramethoxy-6H-dibenzo[a,g]quinolizine, 2,3,9,10-Tetramethoxy-13abeta-berbine, (+)-(R)-Tetrahydropalmatine, (+)-2,3,9,10-Tetramethoxyberbine, (+)-Corydalis B, (R)-Tetrahydropalmatine, (+)-Tetrahydropalmatine Alkaloid, Corydalis B, d-Tetrahydropalmatine
IUPAC navn (13aR)-2,3,9,10-tetramethoxy-6,8,13,13a-tetrahydro-5H-isoquinolino[2,1-b]isoquinoline
SMIL COc1cc2CCN3Cc4c(C[C@@H]3c2cc1OC)ccc(OC)c4OC
Tekniske data
Kemisk navn eller materiale D-Tetrahydropalmatine
Mængde 100 mg
InChI formel InChI=1S/C21H25NO4/c1-23-18-6-5-13-9-17-15-11-20(25-3)19(24-2)10-14(15)7-8-22(17)12-16(13)21(18)26-4/h5-6,10-11,17H,7-9,12H2,1-4H3/t17-/m1/s1
Formel vægt 355.1784