Få mere at vide
D-Mannose-6-phosphate Sodium Salt, TRC
Artikelnummer. 30179610
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179610 5 mg
30179362 10 mg
30178927 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179610 Leverandør Toronto Research Chemicals Leverandørnr. TRCM1850035MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 70442-25-0
Molekylær formel C6H12NaO9P
Molekylvægt (g/mol) 282.12
SMIL [Na+].O[C@H](COP(=O)(O)[O-])[C@@H](O)[C@H](O)[C@H](O)C=O
Tekniske data
Kemisk navn eller materiale D-Mannose-6-phosphate Sodium Salt
Mængde 5 mg
InChI formel InChI=1S/C6H13O9P.Na/c7-1-3(8)5(10)6(11)4(9)2-15-16(12,13)14;/h1,3-6,8-11H,2H2,(H2,12,13,14);/q;+1/p-1/t3-,4-,5-,6-;/m1./s1