Få mere at vide
D,L-Sulforaphane Glutathione, TRC
Artikelnummer. 30136013
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136013 10 mg
30133156 5 mg
30011320 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136013 Leverandør Toronto Research Chemicals Leverandørnr. TRCS69943010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 289711-21-3
Molekylær formel C16 H28 N4 O7 S3
Molekylvægt (g/mol) 484.61
IUPAC navn (2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-(4-methylsulfinylbutylcarbamothioylsulfanyl)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
SMIL CS(=O)CCCCNC(=S)SC[C@H](NC(=O)CC[C@H](N)C(=O)O)C(=O)NCC(=O)O
Tekniske data
Kemisk navn eller materiale D,L-Sulforaphane Glutathione
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C16H28N4O7S3/c1-30(27)7-3-2-6-18-16(28)29-9-11(14(24)19-8-13(22)23)20-12(21)5-4-10(17)15(25)26/h10-11H,2-9,17H2,1H3,(H,18,28)(H,19,24)(H,20,21)(H,22,23)(H,25,26)/t10-,11-,30?/m0/s1
Formel vægt 484.112
Procent renhed >95%