Få mere at vide
D-Idose (0.141M Solution), TRC
Artikelnummer. 17907544
Skift visning
Mængde:
5 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17907544 5 mg
17979844 100 mg
17928794 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17907544 Leverandør Toronto Research Chemicals Leverandørnr. TRCI1750505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5978-95-0
Molekylær formel C6 H12 O6
Molekylvægt (g/mol) 180.1559
Synonym Idose, D- (8CI)
IUPAC navn (2S,3R,4S,5R)-2,3,4,5,6-pentahydroxyhexanal
SMIL OC[C@@H](O)[C@H](O)[C@@H](O)[C@H](O)C=O
Tekniske data
Kemisk navn eller materiale D-Idose
Mængde 5 mg
InChI formel InChI=1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4-,5+,6+/m1/s1
Formel vægt 180.0634
Koncentration eller sammensætning (efter analyt eller komponenter) D-Idose 0.141M