Få mere at vide
Alfa Aesar™ D-Glucal, 97%
Artikelnummer. 15406718
Skift visning
Mængde:
1g
Pakningsstørrelse:
1g
Artikelnummer. Mængde unitSize
15406718 1g 1g
1 options
Artikelnummer. 15406718 Leverandør Alfa Aesar™ Leverandørnr. H60239.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Applications
It is an important building block for both solution- and solid-phase synthesis of oligosaccharides.
Solubility
Soluble in Methanol.
Notes
Store at 2-8°C. Keep away from oxidizing agents. Moisture sensitive.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale D-Glucal
CAS 13265-84-4
Molekylær formel C6H10O4
MDL nummer MFCD09039277
Mængde 1g
Synonym d-glucal, 2r,3s,4r-2-hydroxymethyl-3,4-dihydro-2h-pyran-3,4-diol, glucal, d-arabino-hex-1-enitol, 1,5-anhydro-2-deoxy, 1,5-anhydro-2-deoxy-d-arabino-hex-1-enitol, dgo, pubchem10547, ksc921e7l, 1,2-dideoxy-d-arabino-1-hexenopyranose
InChI nøgle YVECGMZCTULTIS-PBXRRBTRSA-N
Optisk rotation −7° (c=1.9 in Water)
SMIL C1=COC(C(C1O)O)CO
IUPAC navn (2R,3S,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4-diol
Molekylvægt (g/mol) 146.142
PubChem CID 2734736
Formel vægt 146.14
Procent renhed 97%