missing translation for 'onlineSavingsMsg'
Få mere at vide

D-2’-Deoxyribofuranosyl-3-guanylurea(Alpha/Beta-Mixture), TRC

Artikelnummer. 30133570
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133570 25 mg
30133642 50 mg
30006099 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30133570 Leverandør Toronto Research Chemicals Leverandørnr. TRCD25350025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 570410-72-9
Molekylær formel C7H14N4O4
Molekylvægt (g/mol) 218.21
Synonym N-(Aminoiminomethyl)-N'-(2-deoxy-D-erythro-pentofuranosyl)urea;
IUPAC navn N-(Aminoiminomethyl)-N'-(2-deoxy-D-erythro-pentofuranosyl)urea
SMIL O[C@H]1CC(NC(NC(N)=N)=O)O[C@@H]1CO

Specifications

Kemisk navn eller materiale D-2'-Deoxyribofuranosyl-3-guanylurea
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C7H14N4O4/c8-6(9)11-7(14)10-5-1-3(13)4(2-12)15-5/h3-5,12-13H,1-2H2,(H5,8,9,10,11,14)/t3-,4+,5+/m0/s1
Formel vægt 218.1
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.