Få mere at vide
Cysteine-S-sulfate, Monohydrate, TRC
Artikelnummer. 30011458
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011458 250 mg
17972055 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30011458 Leverandør Toronto Research Chemicals Leverandørnr. TRCC997200250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 210110-94-4
Molekylær formel C3 H7 N O5 S2 . H2 O
Molekylvægt (g/mol) 219.24
Synonym L-Cysteine, hydrogen sulfate (ester), monohydrate (9CI)
IUPAC navn (2R)-2-amino-3-sulfosulfanylpropanoic acid;hydrate
SMIL O.N[C@@H](CSS(=O)(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale Cysteine-S-sulfate, Monohydrate
Renhedsgrad noter HPLC
InChI formel InChI=1S/C3H7NO5S2.H2O/c4-2(3(5)6)1-10-11(7,8)9;/h2H,1,4H2,(H,5,6)(H,7,8,9);1H2/t2-;/m0./s1
Mængde 250 mg
Formel vægt 218.9871
Procent renhed >95%