Få mere at vide
Cysteine Protease inhibitor, MedChemExpress
Artikelnummer. 16469857
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16469857 5 mg
16457388 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16469857 Leverandør Medchem Express Leverandørnr. HY17541
MedChemExpress Cysteine Protease inhibitor is an inhibitor of cysteine protease.
Cysteine Protease inhibitor is an inhibitor of cysteine protease.
Tekniske data
Kemisk navn eller materiale Cysteine Protease inhibitor
CAS 921625-62-9
Farve White
Renhedsgrad noter Research
Molekylær formel C18H14N4O
Mængde 5 mg
Opløselighedsinformation DMSO : 50 mg/mL (165.38 mM; Need ultrasonic)
SMIL N#CC1=NC(OC2=CC(C3=CC=C(CN)C=C3)=CC=C2)=CC=N1
Molekylvægt (g/mol) 302.33
Formel vægt 302.33
Procent renhed 96.29%
Grad Research
Fysisk form Solid