Få mere at vide
Cyclizine Lactate, TRC
Artikelnummer. 30367689
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367689 500 mg
30367459 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30367689 Leverandør Toronto Research Chemicals Leverandørnr. TRCC987440500MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5897-19-8
Molekylær formel C21H28N2O3
Molekylvægt (g/mol) 356.46
Synonym 2-Hydroxy-propanoic acid compd. with 1-(diphenylmethyl)-4-methylpiperazine, Lactic acid, compd. with 1-(diphenylmethyl)-4-methylpiperazine, Lactic acid, salt with 1-diphenylmethyl-4-methylpiperazine, 1-(Diphenylmethyl)-4-methyl-piperazine mono(2-hydroxypropanoate), 1-(Diphenylmethyl)-4-methyl-piperazine monolactate, Valoid
IUPAC navn 1-benzhydryl-4-methylpiperazine;2-hydroxypropanoic acid
SMIL CC(O)C(=O)O.CN1CCN(CC1)C(c2ccccc2)c3ccccc3
Tekniske data
Kemisk navn eller materiale Cyclizine Lactate
Mængde 500 mg
InChI formel InChI=1S/C18H22N2.C3H6O3/c1-19-12-14-20(15-13-19)18(16-8-4-2-5-9-16)17-10-6-3-7-11-17;1-2(4)3(5)6/h2-11,18H,12-15H2,1H3;2,4H,1H3,(H,5,6)
Formel vægt 356.21