Få mere at vide
Cyanazine Amide, TRC
Artikelnummer. 30356949
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30356949 100 mg
30365157 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30356949 Leverandør Toronto Research Chemicals Leverandørnr. TRCC953555100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 36576-42-8
Molekylær formel C9H15ClN6O
Molekylvægt (g/mol) 258.71
Synonym 2-[[4-Chloro-6-(ethylamino)-1,3,5-triazin-2-yl]amino]-2-methylpropanamide, 2-Chloro-4-(1-carbamoyl-1-methylethylamino)-6-ethylamino-1,3,5-triazine, 2-Chloro-4-(1-carbamoyl-1-methylethylamino)-6-ethylamino-s-triazine, BV 453, Cyanazine II
IUPAC navn 2-[[4-chloro-6-(ethylamino)-1,3,5-triazin-2-yl]amino]-2-methylpropanamide
SMIL CCNc1nc(Cl)nc(NC(C)(C)C(=O)N)n1
Tekniske data
Kemisk navn eller materiale Cyanazine Amide
Mængde 100 mg
InChI formel InChI=1S/C9H15ClN6O/c1-4-12-7-13-6(10)14-8(15-7)16-9(2,3)5(11)17/h4H2,1-3H3,(H2,11,17)(H2,12,13,14,15,16)
Formel vægt 258.0996