Learn More
Sartorius Cubis™ II Micro Balance, MCE User Interface
BIOPHARMAGREENER_CHOICE
Cubis™ II Micro Balance features fully customizable hardware, software, and connectivity
Brand:  Sartorius MCE3.6P-2S00-F
 View more versions of this product
Product Code. 16407315
  • 197960.00 DKK / Each
Description
  • Monolithical weighing system
  • Integrated climate sensors
  • Individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail
  • State-of-the-art user management
  • Excellent service support
  • Integrated status center
  • Integrated service functions
  • Preventive maintenance on accredited standards
  • TFT touch screen operation with readable display and easy to operate user interface
  • Software Factory installed basic set of essential weighing applications
  • Operation Activated by touch key, touch-free using IR sensor (draft shield M), or gesture sensor (optional) learning capability
Services
10-year warranty
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 USB A ports, 1 USB B port, 1USP C port, 1-9 pin RS 232 connection, 1 ethernet port
Color TFT Touchscreen
Balance, Accessories, Power supply, Manual, Factory test report
Die-cast Aluminum, Plastic PBT, Optiwhite Float Glass and Stainless Steel, Aluminum Trim
±3, 4, 5 μg
Micro Balance
340 mm
129 mm
120/240 V
50/60 Hz
1.1 g, 2.1 g, 3.1 g
1, 2, 5 ÎĽg
±4 μg
Draft shield F: Filtered
3 sec.
10 years
139 mm
6.4, 7.1 kg
100/240 V ±10%
Class I
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Sartorius Cubis™ II Micro Balance, MCE User Interface > MCE user interface; Capacity: 1.1; 2.1; 3.1 g ; Draft shield: F