missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRYZL1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
989.88 DKK - 2678.05 DKK
Specifications
Antigen | CRYZL1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.Specifications
CRYZL1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Non-human Primate, Rat, Human, Mouse | |
O95825, Q921W4 | |
288256, 66609, 9946 | |
CRYZL1 Fusion Protein Ag10419 | |
Primary | |
-20°C | |
CRYZL1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
4P11, CRYZL1, Protein 4P11, QOH 1, Zeta crystallin homolog | |
Cryzl1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CRYZL1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech