Learn More
CRYZL1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16894-1-AP-20UL
 View more versions of this product
Product Code. 16876654
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.
Specifications
Specifications
CRYZL1
Polyclonal
Unconjugated
CRYZL1
4P11, CRYZL1, Protein 4P11, QOH 1, Zeta crystallin homolog
Rabbit
Antigen Affinity Chromatography
RUO
288256, 66609, 9946
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95825, Q921W4
Cryzl1
CRYZL1 Fusion Protein Ag10419
20 ÎĽL
Primary
Non-human Primate, Rat, Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CRYZL1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech > 20 ÎĽL; Unconjugated