Learn More
CRYZL1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16894-1-AP-20UL
 View more versions of this product
Product Code. 16876654
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
This gene encodes a protein that has sequence similarity to zeta crystallin, also known as quinone oxidoreductase. This zeta crystallin-like protein also contains an NAD(P)H binding site. Alternatively spliced transcript variants have been observed but their full-length nature has not been completely determined.
Specifications
Specifications
CRYZL1
Polyclonal
Unconjugated
CRYZL1
4P11, CRYZL1, Protein 4P11, QOH 1, Zeta crystallin homolog
Rabbit
Antigen Affinity Chromatography
RUO
288256, 66609, 9946
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O95825, Q921W4
Cryzl1
CRYZL1 Fusion Protein Ag10419
20 μL
Primary
Non-human Primate, Rat, Human, Mouse
Antibody
IgG
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CRYZL1 Rabbit anti-Human, Mouse, Non-human primate, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated