Learn More
Multiroir™ Crystal Polystyrene Minimax Storage Boxes
Use Multiroir™ Crystal Polystyrene Minimax Storage Boxes for your storage and transport needs. The clear, glass-like polystyrene enables you to view your contents clearly from the outside. The boxes are delivered complete with a separator kit for additional organization of contents.
Brand: Multiroir™ 42009
Additional Details : Weight : 0.45000kg
Description
- Transparent as glass
- Delivered complete with separator kit
- Various capacities
Specifications
Transparent | |
Rectangular | |
1 Each | |
48 mm |
Polystyrene | |
Storage Box | |
315 mm |