missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CREB3L2 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
12030.00 DKK - 19810.00 DKK
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | cAMP responsive element binding protein 3-like 2 |
Gene Alias | BBF2H7,MGC131709,MGC71006 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063485
|
Abnova™
FS0050.100uL |
100 ÎĽL |
12030.00 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16073485
|
Abnova™
FS0050.200uL |
200 ÎĽL |
19810.00 DKK
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
BBF2H7,MGC131709,MGC71006 | |
64764 | |
Liquid |
Tube | |
cAMP responsive element binding protein 3-like 2 | |
CREB3L2 | |
DNA/RNA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Abnova™ CREB3L2 Split FISH Probe