Learn More
Abeba™ Crawler ALU 34555 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Brand: Abeba™ 34555/37
This item is not returnable.
View return policy
Description
- CE EN ISO 20345:2011 SB A, E, FO SRC
- Microfiber
- White
- Aluminum toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3579)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Breathable lining with Silver ions
- Instep area adjustable, with elastic band
- velcro fastening
- Scratch-resistant coating in toe area
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRC | |
Microfiber | |
Slip-resistant | |
34555 |
White | |
37 | |
Polyurethane | |
Aluminum |
Product Content Correction
Product Title
Abeba™ Crawler ALU 34555 Shoes > Size: 37
Spot an opportunity for improvement?Share a Content Correction