Få mere at vide
Cot inhibitor-2, MedChemExpress
Artikelnummer. 16493736
Encompass_Preferred
Skift visning
Mængde:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16493736 25 mg -
16478028 10 mg 10mg
16470336 2 mg -
16460488 5 mg 5mg
16457806 100 mg -
16423957 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Artikelnummer. 16493736 Leverandør Medchem Express Leverandørnr. HY32018
MedChemExpress Cot inhibitor-2 is a potent, selective and orally active cot (Tpl2/MAP3K8) inhibitor with an IC50 of 1.6 nM. Cot inhibitor-2 inhibts TNF-α production in LPS-stimulated human whole blood with an IC50 of 0.3 μM.
Cot inhibitor-2 is a potent, selective and orally active cot (Tpl2/MAP3K8) inhibitor with an IC50 of 1.6 nM. Cot inhibitor-2 inhibts TNF-α production in LPS-stimulated human whole blood with an IC50 of 0.3 μM.
Tekniske data
Kemisk navn eller materiale Cot inhibitor-2
CAS 915363-56-3
Farve Yellow-Green
Renhedsgrad noter Research
Molekylær formel C26H25Cl2FN8
Mængde 25 mg
Opløselighedsinformation DMSO : 40 mg/mL (74.15 mM; Need ultrasonic)
SMIL N#CC1=C(C2=CC(NCC3=CN(N=N3)C4CCN(CC4)CC)=CC(Cl)=C2N=C1)NC5=CC=C(F)C(Cl)=C5
Molekylvægt (g/mol) 539.43
Formel vægt 539.43
Procent renhed 95.14%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease