Få mere at vide
CoraLite™ Plus 488-conjugated MYPT1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
1044.79 DKK - 3358.26 DKK
Tekniske data
| Antigen | MYPT1 |
|---|---|
| Klon | 2A1A9 |
| Koncentration | 1000 μg/mL |
| Applikationer | Immunocytochemistry/Immunofluorescence |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
30233436
|
Proteintech
CL488-66506-20UL |
20 μL |
1044.79 DKK
20µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
30237157
|
Proteintech
CL488-66506-100UL |
100 μL |
3358.26 DKK
100µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
The MYPT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MYPT1. This antibody recognizes human, rat, mouse antigen. The MYPT1 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Tekniske data
| MYPT1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 4659 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 2A1A9 | |
| Immunocytochemistry/Immunofluorescence | |
| CoraLite Plus 488 | |
| Mouse | |
| Human, Rat, Mouse | |
| MBS; MGC133042; MYPT1 | |
| IgG3 | |
| Protein A purification | |
| Conjugated Antibody |