missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CoraLite™ Plus 488-conjugated MYPT1 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66506-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The MYPT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MYPT1. This antibody recognizes human, rat, mouse antigen. The MYPT1 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.Tekniske data
| MYPT1 | |
| Monoclonal | |
| 1000 μg/mL | |
| MBS; MGC133042; MYPT1 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| 115 kDa | |
| Liquid |
| Immunocytochemistry/Immunofluorescence | |
| 2A1A9 | |
| CoraLite Plus 488 | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 4659 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Conjugated Antibody | |
| IgG3 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion