Få mere at vide
Conivaptan Hydrochloride, TRC
Artikelnummer. 30011453
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011453 25 mg
30012165 10 mg
30005162 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011453 Leverandør Toronto Research Chemicals Leverandørnr. TRCC38470025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 168626-94-6
Molekylær formel C32 H26 N4 O2 . Cl H
Molekylvægt (g/mol) 535.04
Synonym N-[4-[(4,5-Dihydro-2-methylimidazo[4,5-d][1]benzazepin-6(1H)-yl)carbonyl]phenyl]-[1,1'-biphenyl]-2-carboxamide Hydrochloride (1:1), [1,1'-Biphenyl]-2-carboxamide, N-[4-[(4,5-dihydro-2-methylimidazo[4,5-d][1]benzazepin-6(1H)-yl)carbonyl]phenyl]-, monohydrochloride (9CI), Conivaptan hydrochloride, Vaprisol, YM 087, conivaptan monohydrochloride
IUPAC navn N-[4-(2-methyl-4,5-dihydro-1H-imidazo[4,5-d][1]benzazepine-6-carbonyl)phenyl]-2-phenylbenzamide;hydrochloride
SMIL Cl.Cc1nc2CCN(C(=O)c3ccc(NC(=O)c4ccccc4c5ccccc5)cc3)c6ccccc6c2[nH]1
Tekniske data
Kemisk navn eller materiale Conivaptan Hydrochloride
Mængde 25 mg
InChI formel InChI=1S/C32H26N4O2.ClH/c1-21-33-28-19-20-36(29-14-8-7-13-27(29)30(28)34-21)32(38)23-15-17-24(18-16-23)35-31(37)26-12-6-5-11-25(26)22-9-3-2-4-10-22;/h2-18H,19-20H2,1H3,(H,33,34)(H,35,37);1H
Formel vægt 534.1823