Learn More
Thermo Scientific™ Complete Manual Staining Assembly 100
Accommodate up to 19 slides with the Thermo Scientific™ Complete Staining Assembly 100, featuring a non-autoclavable glass dish.
185.00 DKK - 309.00 DKK
Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
12667696
View Documents
Thermo Scientific™
101
Staining Dish Cover This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12657696
View Documents
Thermo Scientific™
102
Staining Dish This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Accommodate up to 19 slides with the Thermo Scientific™ Complete Staining Assembly 100, featuring a non-autoclavable glass dish.
Includes:
  • 109 rack/handle
  • 102 dish
  • 101 cover
Staining Rack 109:
Made from stainless steel and holds up to 19 slides. Maximum slide size is 3 x 2 in. (75 x 50mm).
Staining Dish 102:
Made from non-autoclavable glass with inside dimensions of 3.5 x 2.75 x 2.55 in. (90 x 70 x 65mm). Holds up to 250mL of solution with a fully loaded staining rack.
Staining Dish Cover 101:
Made from stainless steel.
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Complete Manual Staining Assembly 100