Få mere at vide
Thermo Scientific™ Complete Manual Staining Assembly 100
Artikelnummer. 12657696 Shop alle Epredia produkter
Produkttype:
Farvning af faddæksel
Farvningsskål
Pakningsstørrelse:
Hver
Artikelnummer. 12657696
Brand: Thermo Scientific™ 102
Accommodate up to 19 slides with the Thermo Scientific™ Complete Staining Assembly 100, featuring a non-autoclavable glass dish.
Accommodate up to 19 slides with the Thermo Scientific™ Complete Staining Assembly 100, featuring a non-autoclavable glass dish.
Includes:
  • 109 rack/handle
  • 102 dish
  • 101 cover
Staining Rack 109:
Made from stainless steel and holds up to 19 slides. Maximum slide size is 3 x 2 in. (75 x 50mm).
Staining Dish 102:
Made from non-autoclavable glass with inside dimensions of 3.5 x 2.75 x 2.55 in. (90 x 70 x 65mm). Holds up to 250mL of solution with a fully loaded staining rack.
Staining Dish Cover 101:
Made from stainless steel.
TRUSTED_SUSTAINABILITY
Tekniske data
Dybde (engelsk) Indvendig 2.55 in.
Dybde (metrisk) Indvendig 65 mm
Fad Materiale Non-autoclavable Glass
Længde (engelsk) Indvendig 3.5 in.
Længde (metrisk) Indvendig 90 mm
Bredde (engelsk) Indvendig 2.75 in.
Bredde (metrisk) Indvendig 70 mm
Produkttype Farvningsskål
Løsningskapacitet 250 mL