missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Novus Biologicals™ Complement Component C2 Antibody (ARGX-117) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28215-100ug
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Complement Component C2 Monoclonal antibody specifically detects Complement Component C2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| Complement Component C2 | |
| Monoclonal | |
| Unconjugated | |
| P06681 | |
| Human | |
| Protein A purified | |
| RUO | |
| 717 | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| ARGX-117 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| C3/C5 convertase, CO2, complement C2, complement component 2, complement component C2, DKFZp779M0311, EC 3.4.21, EC 3.4.21.43 | |
| Complement C2 | |
| 100 μg | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion